PolyTack Premium Mat, Connecticut Cleanroom CORP
Supplier: Connecticut Cleanroom CORP
P-104G P-102W P-107G P-101G P-101W P-104B P-105B P-102B P-105W P-111W P-104W P-102G P-107B P-101B P-112B P-101C P-105G
53333-208CS 615.95 CAD
53333-208 53333-206 53333-204 53333-258 53333-202 53333-268 53333-212 53333-256 53333-002 53333-200 53333-254 53333-210 53333-000 53333-264 53333-252 53333-262 53333-250
PolyTack Premium Mat, Connecticut Cleanroom CORP
Floor Mats Traction and Anti-Fatigue Floor Mats
Made from an incinerable polyethylene film, mats remove foot borne particulates before entering the cleanroom
Seamless one-piece construction. Numbered tabs provide easy identification and sheet separation. Easy mobility for carts and wheeled machinery. Polyethylene film contains a special anti-microbial agent providing long-lasting protection against bacterial growth.
Four mats per case, with each mat consisting of 30 layers.