Sample Vials, Clear, PTFE Lined Caps, Chemglass
Supplier: Chemglass
CG-4900-01 CG-4900-02 CG-4900-03 CG-4900-04 CG-4900-05 CG-4900-06
33502-054EA 588.02 CAD
33502-054 33502-056 33502-058 33502-060 33502-062 33502-064
Sample Vials, Clear, PTFE Lined Caps, Chemglass
Vials
Sample vials are made from Type 1 Borosilicate glass.
  • Full height partitions minimize breakage
  • Solid PTFE lined caps
Packaging: Packaged in convenient LAB-PACS of 144 pieces.