Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Pure-Fit® SIB®
PF135375PPAF PFMS625PVDF PFX375PVDF PFMS250PPAF PF135500PPAF PFLS500PVDF PFX375PPAF PFT187PPAF PFY312PPAF PFT125JAW360 PFR250X375PVDF PFR062X250PVDF PFT250PPAF PFMS500PPAF PFMS500PVDF PFMS375PVDF PFY375PVDF PFMS125PPAF PF135500PVDF PFE375PVDF PFC125PPAF PFR250X375PPAF PFC312PVDF PFX250PVDF PFMS375PPAF PFC375PVDF PFX250PPAF PFT500PPAF PFR187X312PPAF PFC250PVDF PFT500PVDF PFE250PVDF PFR125X187PPAF PFR187X375PVDF PFT375PVDF PFE375PPAF PFT750PPAF PFMS125PVDF PFLS500PPAF PFE500PPAF 75830-908 PFLS250PPAF PFT187JAW360 PFLS375PPAF PFR250X500PVDF PFR250X500PPAF PFC312PPAF PFY250PPAF PF135375PVDF PFC187PPAF PFR187X375PPAF PFR375X500PPAF PFMS250PVDF 75830-878 PFT125PPAF PFY500PVDF PFT187PVDF PFY125PVDF PFR375X500PVDF PFE250PPAF PFT125PVDF PFT135312 PFX500PVDF PFC250PPAF PFR187X312PVDF PFLS375PVDF PFX125PPAF PFLS750PPAF PF135250PVDF PFY250PVDF PFLS750PVDF PFT062JAW360 PFT135125 PFX500PPAF PFR062X125PVDF PFY500PPAF PFT312PVDF PFT750PVDF PFX125PVDF PFE500PVDF PFT312JAW360 PFR125X187PVDF PFT250PVDF PFC125PVDF PFLS250PVDF PFY312PVDF PFR125X250PVDF PFC500PVDF PFC500PPAF PFX187PVDF PFC375PPAF PFY375PPAF PFT312PPAF PFT135062 PFR062X250PPAF PFT135187 PFC187PVDF PFX187PPAF PFMS625PPAF PFR125X250PPAF PF135250PPAF PFR062X125PPAF
75830-928BG 540.04 CAD
75830-928 75830-926 75830-924 75830-922 75830-888 75830-920 75830-886 75830-896 75830-894 75830-892 75830-890 75831-046 75831-048 75831-042 75830-918 75831-044 75830-916 75830-914 75831-040 75830-912 75830-878 75830-910 75830-998 75830-876 75830-996 75830-884 75830-882 75830-880 75831-038 75831-034 75831-036 75830-908 75831-030 75830-906 75831-032 75830-904 75830-902 75830-900 75830-988 75830-986 75830-874 75830-994 75830-872 75830-992 75830-870 75830-990 75831-028 75831-024 75831-026 75831-020 75831-022 75830-978 75830-976 75830-974 75830-984 75830-982 75830-980 75831-016 75831-018 75831-012 75831-014 75831-096 75831-098 75831-010 75831-092 75830-968 75831-094 75830-966 75830-964 75831-090 75830-972 75830-970 75831-006 75831-008 75831-002 75831-004 75831-086 75831-088 75831-000 75831-120 75831-082 75831-084 75831-080 75830-962 75831-116 75831-118 75831-112 75831-078 75831-114 75831-074 75831-110 75831-076 75831-070 75831-072 75831-108 75831-104 75831-106 75831-068 75831-100 75831-102 75831-066 75830-898
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Fittings and Connectors
Pure-Fit SIB is a smooth inner bore (SIB) hose-barb connector that provides a seamless transition between tubing and fittings throughout the fluid path.
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilizes the patented BarbLock retention and seal system
  • Compatible with Sani-Tech, C-Flex and PharMed flexible tubing
Available in both animal-free polypropylene (PP) material, which is gamma stable and polyvinylidene fluoride (PVDF) material, which is both autoclavable and gamma stable.
Typical areas of applications include biopharmaceutical manufacturing, media hydration, filtration, cell culture/harvest, product sampling and downstream activities.
Certifications: Meets all USP Class VI and ISO criteria.