Pure-Fit® TCL Large Tube Clamps, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
PFTCL150 PFTCL250
75831-060BG 2418.6 CAD
75831-060 75831-062
Pure-Fit® TCL Large Tube Clamps, Saint-Gobain Performance Plastics
Clamps Tubing and Sanitary Clamps
Pure-Fit® TCL large tube clamps are designed to accommodate large diameter and thick wall tubing applications. They are compatible with standard, braided (reinforced) and multilayer tubing. Smooth edges and corners help prevent punctures and ruptures.
  • Provides complete fluid stoppage and can be utilized in the flow position
  • Features an integral bore hole for absolute lock-out capabilities
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Fully autoclavable and sterilizable
Constructed from animal derivative-free and BPA-free PVDF and available in two sizes to accommodate a wide range of thick wall tubing.
Areas of use include biopharmaceutical manufacturing, pharmaceutical processes, peristaltic pump sets, drug delivery & discovery, medical systems, laboratory functions and assemblies and tubing sets.
Certifications: Meets all USP Class VI criteria.