Pure-Fit® TC Standard Size Tube Clamps, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
PFTC375 PFTC375-250 PFTC750PVDF-UA PFTC375PVDF-UA PFTC750 PFTC750-250
75831-050BG 279.69 CAD
75831-050 75831-052 75831-054 75831-056 75831-058 75831-122 75831-124 75831-126 75831-128 75831-130 75831-132 75831-134 75831-136
Pure-Fit® TC Standard Size Tube Clamps, Saint-Gobain Performance Plastics
Clamps Tubing and Sanitary Clamps
Pure-Fit® TC tube clamps are ideal for use in the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; and a side release mechanism that facilitates secure, single-handed installation and operation.
  • Integral bore hole for lock-out capabilities
  • Provides complete fluid stoppage
  • Internal ratchet mechanism meters fluid flow
  • Can be installed on or over tubing or finished assemblies
More Product Information