Pure-Fit® Secure Aseptic Connectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
PFSC250-IHM PFSC250-OHM PFSC375-CAM PFSC375-IHM PFSC375-OHM PFSC500-CAM PFSC500-IHM PFSC500-OHM PFSC750-CAM PFSC750-IHM PFSC750-OHM PFSC1.00-CAM PFSC1.00-IHM PFSC1.00-OHM
75830-650BG 4016.3 CAD
75830-650 75830-652 75830-654 75830-656 75830-658 75830-660 75830-662 75830-664 75830-858 75830-860 75830-862 75830-864 75830-866 75830-868
Pure-Fit® Secure Aseptic Connectors, Saint-Gobain Performance Plastics
Fittings and Connectors
Pure Fit® secure connectors made of fully characterized, animal derivative-free and BPA-free materials that makes completely aseptic connections possible outside the cleanroom environment.
  • Silicone valve design leaves no gaps or residue
  • Easy-to-see laser-etched visual indicators confirm proper engagement
  • Can be sterilized via gamma irradiation or autoclaving
Pure-Fit® SIB® technology on barb connections ensures a completely smooth fluid path
Available in interchangeable 3/4" and 1'' sizes for greater flow volume and sizes 1/4", 3/8", 1/2" for step-More Product Information