Sani-Tech® STHT®-C Silicone Tubing
Supplier: Saint Gobain Performance Plastics
STHT-C-030-2 STHT-C-062-3 STHT-C-078-4 STHT-C-093-1 STHT-C-125-2F STHT-C-187-3F STHT-C-750-1 STHT-C-875-4 STHT-C-750-3 STHT-C-375-2 STHT-C-375-6
75829-372PK 133.75 CAD
75829-372 75829-386 75829-388 75829-390 75829-392 75829-394 75829-396 75829-398 75829-400 75829-402 75829-404 75829-406 75829-408 75829-410 75829-412 75829-414 75829-416 75829-418 75829-420 75829-422 75829-424 75829-426 75829-428 75829-430 75829-432 75829-434 75829-436 75829-438 75829-440 75829-442 75829-444 75829-446 75829-448 75829-450 75829-452 75829-454 75829-456 75829-458 75829-460 75829-462 75829-464 75829-468 75829-470 75829-472 75829-474 75829-476 75829-478 75829-480 75829-482 75829-484 75829-486 75829-488 75829-490 75829-492 75829-494 75829-496 75829-498 75829-500 75829-502 75829-504 75829-506 75829-508 75829-510 75829-512 75829-514 75829-516 75829-518 75829-520 75829-522 75829-524 75829-526 75829-528 75829-530 75829-532 75829-534 75829-536 75829-538 75829-540 75829-542 75829-544 75829-546 75829-548 75829-550 75829-554 75829-556 75829-558 75829-560 75829-562 75829-564 75829-566 75829-568 75829-570 75829-572 75829-574 75829-576 75829-578 75829-580 75829-582 75829-584 75829-586 75829-588 75829-590 75829-592 75829-594 75829-596 75829-598 75829-600 75829-602 75829-604 75829-606 75829-608 75829-610 75829-612 75829-614 75829-616 75829-618 75829-620 75829-622 75829-624 75829-626 75829-628 75829-630 75829-632 75829-634 75829-636 75829-638 75829-640 75829-642 75829-644 75829-646 75829-648 75829-650 75829-652 75829-654 75829-656 75829-658 75829-660 75829-662 75829-664 75829-666 75829-668 75829-670 75829-672 75829-674 75829-676 75829-678 75829-680 75829-682 75829-684 75829-686 75829-688 75829-690 75829-692 75829-694 75829-696 75829-698 75829-700 75829-702 75829-704 75829-706 75829-708 75829-710 75829-712 75829-714 75829-896 75829-898 75829-900 75829-902 75829-904 75829-906 75829-908 75829-910 75829-912 75829-914 75829-916 75829-918 75829-920 75829-922 75829-924 75829-926 75829-928 75829-930 75829-932 75829-934 75830-106 75830-108 75830-110 75830-112 75830-114 75830-116 75830-118 75830-120 75830-122 75830-124 75830-126 75830-128 75830-130 75830-132 75830-134 75830-136 75830-138 75830-140 75830-142 75830-326 75830-328 75830-330
Sani-Tech® STHT®-C Silicone Tubing
Tubing Transfer Tubing
Clear platinum-cured non-reinforced silicone tubing made from ultra-pure, biopharmaceutical grade silicone.
  • Ultra-flexible, autoclavable and sterilizable
  • Unsurpassed surface smoothness
  • Imparts no taste or odors
  • Permanently laser marked with part number and lot code for total product traceability
  • Packages are externally labelled on both the bag and the crush-proof box
Sani-Tech® STHT®-C silicone tubing resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack.
Sani-Tech® STHT®-C siliconeMore Product Information