PROSAT® Sterile Low Endotoxin Knitted Wipes, Contec®
Supplier: Contec
PSWE0002 PSWE0001 PSWE0003
52428-882CS 3459.13 CAD
52428-882 75779-606 75833-990
PROSAT® Sterile Low Endotoxin Knitted Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
PROSAT® Sterile Polynit Heatseal LE Wipes are made of 100% knitted polyester Polynit Heatseal, presaturated with a blend of 70% isopropanol and 30% water for injection (WFI). Each lot is tested before release and low endotoxin certified to less than 1 EU/wipe.
  • Recommended for Grade A/B or ISO Class 5 environments (product contact areas)
  • 100% knitted polyester fabric results in very low levels of particles and fibers
  • Laser sealed edges reduce fiber generation from the edge of the wipe
  • Guaranteed endotoxin levels of 1EU/device make them ideal for product
More Product Information