Mag-Bind® Plant DNA DS 96 Kit, Omega
Supplier: OMEGA BIO-TEK CA
The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content.
- Up to 50 mg wet tissue can be processed in less than one hour
- Increase yields and provide high-quality DNA
- Eliminates the need for chloroform extractions
The Mag-Bind® Plant DNA DS 96 Kit allows rapid and reliable isolation of high-quality genomic DNA from plants and other tissues that are particularly difficult to lyse or very high in polysaccharide content. The lysis and binding buffers are specifically designed to minimize co-purification of polysaccharides and polyphenols. Up 96 samples of 50 mg wet tissue (or 15 mg dry tissue) can be processed in parallel in less than one hour. The system combines CTAB-based lysis, which eliminates the need for organic solvents, with the convenience of Mag-Bind® Particles to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. This kit is designed for manual or fully automated high throughput preparation of genomic, chloroplast, and mitochondrial DNA. Purified DNA is suitable for PCR, restriction digestion, next-generation sequencing, and hybridization applications. There are no organic extractions thereby reducing consumables and decreasing hands-on time to allow multiple samples to be processed in parallel.
Protocols are available for the following automated platforms:
Hamilton Microlab® STAR
Hamilton Microlab® NIMBUS
KingFisher™, BioSprint®, and MagMAX® 96