E-Z 96® Plant DNA DS Kit, Omega

Supplier: OMEGA BIO-TEK CA

D1411-00 D1411-01
CA75818-248EA 460.57 CAD
CA75818-248 CA75818-250
E-Z 96® Plant DNA DS Kit, Omega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content.


  • Up to 50 mg wet tissue can be processed in less than one hour
  • Increase yields and provide high-quality DNA
  • Eliminates the need for chloroform extractions


The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content. Up to 50 mg wet tissue can be processed in less than one hour. The system combines the reversible nucleic acid-binding properties of the HiBind® matrix with the speed and versatility of spin column technology to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates.  Purified DNA is suitable for PCR, restriction digestion, and hybridization applications. This procedure relies on the well-established properties of the cationic detergent, cetyltrimethyl ammonium bromide (CTAB), in conjunction with the unique binding system to increase yields and provide high-quality DNA.  The system eliminates the need for chloroform extractions traditionally associated with CTAB-based lysis methods. Samples are homogenized and lysed in a high salt buffer containing CTAB, binding conditions are adjusted, and DNA is purified using a E-Z 96 DNA Plate. Salts, proteins, and other contaminants are removed to yield high-quality genomic DNA suitable for downstream applications such as endonuclease digestion, thermal cycle amplification, and hybridization applications.

Order Now

SPECIFICATIONS

Description E-Z 96® Plant DNA DS Kit
Environmentally Preferable
Application
Target molecule DNA
Refine target molecule Total DNA
Sample type Plant samples
Method / Format Multiwell plate
Prep size Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR