Boreal Research Microscope Series
Supplier: National Optical
168-SP 168-ASC 169-SP 169-ASC
470222-444EA 1494.7 CAD
470222-444 470222-446 470222-448 470222-450
Boreal Research Microscope Series
Microscopes
This microscope is ideal for advanced studies and laboratory use.
  • Choice of Objective Lenses: ASC or Semi-Plan
  • LED Illumination
  • Large, Built-In, Low-Profile, Rack-Less Stage
  • Manufacturer's Lifetime Warranty
  • Superb Optical Clarity
  • Color-Coded Objectives
The new Research series incorporates a wide range of features designed for university and laboratory use. Superb optical clarity and reliable mechanical engineering assure years of top performance.
Two grades of objective lenses are available, both of which are parfocal, parcentered, and color coded.
Ordering information: This product includes a manufacturer's warranty.