Econo™ Sterile Hoff Towel Clamps, Sklar
Supplier: Sklar
96-2564 96-2565
89023-874CS 1569.57 CAD
89023-874 89023-876
Econo™ Sterile Hoff Towel Clamps, Sklar
Forceps
These are non-perforating clamps used to secure instruments and towels to the drape surrounding a surgical site.
  • Shanks and handle are flat so they can rest on the patient
  • Made of stainless steel
The tip of this towel clamp are round with a cross-serrated surface so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured.
This clamp should never be used on skin. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.