Accessories for Corning® Microcarriers, Corning
Supplier: Corning
4626 4627 4628 4629
10002-716CS 293.11 CAD
10002-716 10002-718 10002-720 10002-722
Accessories for Corning® Microcarriers, Corning
Beads Microcarriers
These polypropylene aseptic transfer screw caps prevents adventitious contamination in for microcarriers bottles during bioreactor loading.
  • Chemically resistant and heat sealable flexible tubing
  • Tubing length with connector 30" (76.2 cm)
  • Disposable and sterile (10⁻⁶ SAL)
Packaging: These caps are individually double bagged.