Microbiological Incubators, Avantgarde.line BF Series, BINDER
Supplier: Binder
9010-0316 9010-0320 9010-0314
75783-546EA 6355.08 CAD
75783-546 75783-548 75783-550 75783-612 75783-614 75783-616
Microbiological Incubators, Avantgarde.line BF Series, BINDER
Incubators Standard Incubators
BINDER microbiological incubators are ideal for research, production and quality assurance.
  • APT.Line™ preheating chamber
  • Microprocessor PID controller
These incubators feature the APT.Line™ preheating chamber for reliable temperature accuracy and uniformity in applications requiring constant, reproducible incubation conditions
Units have an inner tempered safety glass door, corrosion resistant stainless steel chamber and two chrome plated racks. They also carry a Class 3.1 independent temperature safety device with visual and audible temperatureMore Product Information