Standard-Incubators with Natural Convection, Avantgarde.line BD Series, BINDER
Supplier: Binder
9010-0324 9010-0326 9010-0330
75783-554EA 3584.39 CAD
75783-554 75783-558 75783-618
Standard-Incubators with Natural Convection, Avantgarde.line BD Series, BINDER
Incubators Standard Incubators
The BD Avantgarde.Line series is specially designed for gentle incubation of organisms, as well as conditioning of heatsensitive media. The design of the units and the homogeneous temperature distribution ensure the very best results.
  • Excellent temporal and spatial temperature accuracy with high process reliability
  • Disinfection routine of 100 °C
  • High temperature accuracy with APT.line™ technology
  • Controller with LCD display
  • Electromechanical control of the exhaust air flap
  • Ergonomic handle design
  • USB port for recording data
Features inner door made ofMore Product Information