Drying/Heating Ovens with Natural Convection, Avantgarde.line ED Series, BINDER
Supplier: Binder
9010-0334 9010-0336 9010-0340
75783-622EA 2875.76 CAD
75783-622 75783-626 75783-630
Drying/Heating Ovens with Natural Convection, Avantgarde.line ED Series, BINDER
Ovens Standard Ovens
BINDER Avantgarde.Line ED Drying ovens are ideal for sterilization, drying, and thermal storage tasks which do not require high drying rates or special time parameters.
  • Ovens provide reliable, reproducible, quick task results
  • Easy to clean
BINDER ovens provide fast uniform drying for drying and sterilization applications. The Advanced Preheating Technology (APT.line™) provides temperature accuracy of 1°C with a wide temperature range from 5°C to 15°C above ambient to 300 °C (depending on the model size). With the natural convection and high rate ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable