Mixer Vortex Genie® 2 and 2T, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71840 71840-T
102091-234EA 1121.59 CAD
102091-234 102097-722 102097-732 102097-734
Mixer Vortex Genie® 2 and 2T, Electron Microscopy Sciences
Shakers Vortex Mixers
Versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision.
This Vortex-Genie has unmatched reliability and multi-task versatility.
The 2T has timer function, both Touch and Hands-Free mode automatically turns the unit off after set time. Touch Mode: 1 to 60 seconds. Hands-Free (Switch activated Start/Stop) Mode: 1 to 60 minutes, or continuous un-timed mode. Unit automatically stops at set time – in Touch Mode or Hands-Free (switch activated Start/Stop) Mode. Accuracy ±5%. Repeatability ±1%.
It features shaking or vortexing action. Variable speed control allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing to choose the correct action for application. A broad range of attachments of various shapes, sizes, and materials allows for the mixing of almost all common tubes or container for hands-free or touch operation. A 3-position power switch lets to select continuous action or "Touch On" control. With the true "Hands- Free" operation of continuous action, lab operators can greatly increase lab efficiency. Tough metal housing combined with "just enough weight", provides a stable platform for all types of mixing.
Accessories information: Snap on these attachments for new ways to use your Vortex-Genie. Note that items marked with an asterisk (*) are included with Vortex-Genie 2 and Vortex-Genie 2T.
Delivery information: All units include a 3" Platform, pop-off cup, cast metal base with rubber feet of base dimensions: 165×122×165 mm (6.5×4.8×6.5").