Centrifuge Tubes
Supplier: TWD Scientific
BX10-0151 BX10-0152 BX10-0501 BX10-0502
75799-886CS 402.06 CAD
75799-886 75799-888 75799-890 75799-892 75799-894 75799-898 75799-900
Centrifuge Tubes
Tubes Centrifuge Tubes
Centrifuge tubes feature conical bottoms for superior separation and pelleting.
  • Gamma radiation sterilized and non-pyrogenic
  • 15 and 50 ml conical tubes
  • Autoclavable
  • Graduated
  • Tubes withstand centrifugation of 8400 RCF
  • Medical-grade polypropylene construction
  • Transparent wall permits easy viewing of tube content
Tubes are supplied with either a Plug-Seal cap or a Flat-Top cap.