TWD Barcoded Autosampler Vials
Supplier: TWD Scientific
GV1232A-P-2DK GV1232C-P-2DK GV1232A-P-2DLE GV1232C-P-2DLE
75780-316CS 191.46 CAD
75780-316 75780-318 75780-320 75780-342 75780-344
TWD Barcoded Autosampler Vials
Vials
TWD Barcoded Autosampler Vials with 2D barcode.
  • Vials feature writing patch and graduations
  • Glass construction
  • Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
Delivery information: Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.