Biox Cardboard Freezer Boxes
Supplier: TWD Scientific
BX90-5281 BX90-5200
75780-334CS 545.56 CAD
75780-334 75780-336
Biox Cardboard Freezer Boxes
Boxes Cooling/Freezing Boxes and Bags
Premium Cardboard Freezer Boxes with Cryo-Color coating.
  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250°F)
  • Accommodates 0.5mL, 1.5mL, or 2.0mL microtubes
  • Suitable for liquid nitrogen storage
Assorted colors for easy identification and water resistant coating improved durability.