Gordon Brush® Metal Free Tube Brushes
Supplier: Gordon Brush
732518 730718 732018 731518 741048 721824 721812 721524 710518 711212 720718 710618 720824 710624 740536 711012 721224 722524 710324 732524 721024 720712 721512 731712 731524 710512 731512 711024 730712 741024 740724 721818 710824 710612 732512 720812 710718 740748 722518 710212 710818 731018 731012 710312 740736 720818 733018 722018 733012 731224 711018 710318 730724 741036 710224 721218 722512 710724 721012 721518 733024 740524 731218 722024 711224 731724 731212 732012 732024 710712 731718 722012 720724 731024 721212 710524 710218 740548 721018 710812 711218
75816-580PK 934.65 CAD
75816-580 75816-536 75816-658 75816-614 75816-578 75816-534 75816-612 75816-656 75816-618 75816-538 75816-616 75816-572 75816-650 75816-570 75816-532 75816-576 75816-610 75816-654 75816-530 75816-574 75816-652 75816-590 75816-624 75816-504 75816-548 75816-622 75816-666 75816-502 75816-546 75816-628 75816-508 75816-626 75816-506 75816-660 75816-584 75816-540 75816-582 75816-620 75816-664 75816-588 75816-544 75816-662 75816-542 75816-586 75816-514 75816-558 75816-636 75816-556 75816-512 75816-518 75816-516 75816-638 75816-550 75816-594 75816-592 75816-598 75816-554 75816-510 75816-552 75816-596 75816-630 75816-608 75816-646 75816-602 75816-526 75816-600 75816-644 75816-568 75816-524 75816-606 75816-604 75816-648 75816-528 75816-562 75816-560 75816-642 75816-522 75816-566 75816-640 75816-564 75816-520
Gordon Brush® Metal Free Tube Brushes
Brushes
Metal Free Tube Brushes are manufactured with 100% food grade polypropylene handle and bristles. Polypropylene is resistant to acids, alkalis, alcohols, and many petroleum distillates. These brushes can be used to clean the inside of straight or curved glassware, tubes, pipes, valves, and fittings. They are ideally suited for moderate to aggressive scrubbing. Brushes will not scratch most surfaces, and the fused bristles help minimize the growth of bacteria. Available in white only.
  • 100% food grade polypropylene handle and bristles
  • Designs available to clean straight or curved glassware
  • Resistant to acids, alkali, alcohol, and petroleum distillates
  • Ideal for moderate to aggressive scrubbing
  • Fused bristles minimize bacteria growth
Metal Free Tube Brushes are available in four distinct designs to accommodate a wide variety of laboratory cleaning requirements.
The Flexible Series brushes feature a flexible stem specifically designed to access the interior of straight or curved pipes and tubes. Standard Series brushes are equipped with a semi-flexible stem designed to clean the inside of pipes and tubes with a gradual bend. Rigid Series brushes feature a strong and non-flexible stem designed for use inside straight pipes and tubes. Double Leader Series brushes feature a push/pull-through design, with a 20.3cm (8") brush head and a ball on each end.