SP Wilmad-LabGlass Extension Clamps, Three Prong, SP Industries
Supplier: SP Industries
LG-4637-108 LG-4637-110
14008-154EA 95.59 CAD
14008-154 14008-156 14008-158 14008-160 14008-162 14008-164
SP Wilmad-LabGlass Extension Clamps, Three Prong, SP Industries
Clamps General Purpose Clamps
These extension clamps are three-pronged and come in single or dual adjustment - both available in three sizes to provide a wide range of jaw openings.
  • Three-pronged
  • Wide range of jaw openings
  • Hold irregular shapes easily
The three-jaw configuration secures various lab apparatus and holds irregular shapes easily. Long, seamless brass tubing attaches clamping heads securely and offers easy positioning and depth adjustment, even in the deepest fume hoods. High tensile strength zinc alloy construction; exposed surfaces are nickle plated for maximum corrosion resistance.