HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5% (by titrimetric analysis on dry substance), white powder

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

00174-10KG 00174-1KG 00174-250G 00174-25G
CH00174-10KGEA 2297.29 CAD
CH00174-10KG CH00174-1KG CH00174-250G CH00174-25G
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5% (by titrimetric analysis on dry substance), white powder
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

HEPES is used as a hydrogen ion buffer, which can control the constant pH range for long term.


  • Non-toxic to cell

Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
MDL Number: MFCD00006158
CAS Number: 7365-45-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder
Solubility (0.1M in Water) Clear and complete
pH (5% Solution in H2O) 5.0 - 6.5
pKa (25 °C) 7.35 - 7.75
Loss on Drying ≤0.5%
Ash ≤0.3%
Assay by titration (dry basis) ≥99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR