20X PBS (Phosphate Buffered Saline), pH 7.4
Supplier: GROWCELLS
MRGF-6395 MRGF-6396 MRGF-6395-010L MRGF-6395-020Q
75801-006CS 1971.98 CAD
75801-006 75801-008 75801-010 75801-014 75801-016
20X PBS (Phosphate Buffered Saline), pH 7.4
Buffers Cell Culture Buffers
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 20X concentration to suit a variety of working dilution for any biological need.