PBS (Phosphate Buffered Saline), 10X, pH 7.4, GrowCells
Supplier: GROWCELLS
MRGF-6235 MRGF-6236 MRGF-6235-010L MRGF-6235-010Q MRGF-6235-020Q MRGF-6235-55GL
75800-994CS 1277.78 CAD
75800-994 75800-996 75800-998 75801-000 75801-002 75801-004
PBS (Phosphate Buffered Saline), 10X, pH 7.4, GrowCells
Buffers Cell Culture Buffers
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 10X concentration to suit a variety of working dilution for any biological need.