1X PBS (Phosphate Buffered Saline), pH 7.4
Supplier: GROWCELLS
MRGF-6230 MRGF-6231 MRGF-6230-010L MRGF-6230-010Q MRGF-6230-020Q
75800-982CS 1215 CAD
75800-982 75800-984 75800-986 75800-988 75800-990 75800-992
1X PBS (Phosphate Buffered Saline), pH 7.4
Buffers Cell Culture Buffers
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 1X concentration to suit a variety of working dilution for any biological need.