10X TBE (Tris-Borate EDTA) pH 8.3
Supplier: GROWCELLS
MRGF-4215 MRGF-4216 MRGF4216010L MRGF4216010Q MRGF4216020Q
75800-946CS 1344.66 CAD
75800-946 75800-948 75800-950 75800-952 75800-954 75800-956
10X TBE (Tris-Borate EDTA) pH 8.3
Buffers Electrophoresis Buffers Running Buffers
TBE is the most commonly used buffer for DNA and RNA electrophoresis
Since borate is a strong inhibitor of enzymes, TBE is recommended for DNA/RNA sizing, not for recovery of DNA for enzymatic applications. Routinely used for DNA automated sequencing gels. Used both as a running buffer and gel preparation buffer. Recommended for use with fragments smaller than 1500 bp.