50X TAE (Tris-Acetate EDTA), pH 8.3
Supplier: GROWCELLS
MRGF-4210 MRGF-4211 MRGF-4210-010Q MRGF-4210-020Q
75800-934CS 3095.06 CAD
75800-934 75800-936 75800-938 75800-940 75800-942 75800-944
50X TAE (Tris-Acetate EDTA), pH 8.3
Buffers Electrophoresis Buffers Running Buffers
TAE buffer is used in agarose gel electrophoresis and Polyacrylamide gels both as a buffer and a component of a gel
Can be used for both genomic and large supercoiled DNA. TAE buffer has a relatively low buffering capacity therefore buffer should be replaced periodicly during long electrophoresis runs. Offered in different concentration for all common biological applications.
Order Now
SPECIFICATIONS
Environmentally Preferable