EDTA, pH 8.0, 0.5M
Supplier: GROWCELLS
MRGF-1202 MRGF-1203 MRGF-1203-010L MRGF-1203-010Q
75800-466CS 3100.32 CAD
75800-466 75800-468 75800-470 75800-472 75800-474 75800-920
EDTA, pH 8.0, 0.5M
Buffers Biological Buffers
Ethylenediaminetetraacetic acid is a polyamino carboxylic acid, commonly used to deactivate metal dependent enzymes, or as a chelating agent that binds to metal ions
Suitable for RNA storage. Offered in different molar concentration for all common biological applications.