3M Sodium Acetate Solution, pH 5.5
Supplier: GROWCELLS
MRGE-5265 MRGE-5266-020Q
75800-476CS 3252.49 CAD
75800-476 75800-892 75800-894 75800-896
3M Sodium Acetate Solution, pH 5.5
Buffers Biological Buffers
3M Sodium acetate.
Ready-to-use sterile solution made with 18 megOhm water, 0.2 µm sterile filtered, pH adjusted to 5.5 with glacial acetic acid. Ideal for DNA purification.