Quidel QuickVue® In-Line® One Step Strep A, Hardy Diagnostics
75813-938EA 317.8 CAD
75813-938
Quidel QuickVue® In-Line® One Step Strep A, Hardy Diagnostics
Assays Immunological Assays
QuickVue® In-Line® One Step Strep A, rapid detection of Group A streptococcal antigen directly from patient throat swab specimens.
  • Allows testing to take place at the point of care
  • Two-color results give you clear readability
  • Dependable results in five minutes or less
A true one-step test and is the first Strep A test waived under CLIA "88. The unique in-line extraction and rapid procedure allow testing to take place at the point of care. Two-color results give you clear readability, and the proven accuracy of the QuickVue In-Line Strep A test provides dependable results in five minutes or less and 96% overall agreement with SBA culture. Individually packaged for convenient single-use testing and disposal. Positive and negative external controls included in the kit.