VWR® Rochester Ocshner Kocher Rat Tooth Forceps
Supplier: VWR International
10806-098 10806-100 10806-102 10806-104 10806-106 10806-158 10806-160 10806-162
10806-098EA 62.11 CAD
10806-098 10806-100 10806-102 10806-104 10806-106 10806-158 10806-160 10806-162
VWR® Rochester Ocshner Kocher Rat Tooth Forceps
Forceps
VWR® Rochester Ocshner Kocher Rat Tooth Forceps are constructed of surgical grade stainless steel
  • Passivated
  • Rockwell Hardness (HRC): 40–45
  • Box-lock joint
  • Surgical grade
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are available in curved or straight formats, in a variety of lengths. All forceps feature serrated jaws with rat tooth tips in a 1 x 2 teeth configuration. Forceps are fully autoclavable, and can be sterilized by EtO/gamma radiation.