VWR® Pean Hemostatic Forceps
Supplier: VWR International
10806-128 10806-130 10806-134 10806-182 10806-184 10806-186 10806-188 10806-190 10806-192 10806-194 10806-196 10806-198 10806-200 10806-202 10806-204 10806-206 10806-208 10806-210
10806-128EA 31.71 CAD
10806-128 10806-130 10806-134 10806-182 10806-184 10806-186 10806-188 10806-190 10806-192 10806-194 10806-196 10806-198 10806-200 10806-202 10806-204 10806-206 10806-208 10806-210
VWR® Pean Hemostatic Forceps
Forceps
VWR® Pean Hemostatic Forceps are constructed of surgical grade stainless steel
  • Passivated
  • Box-lock joint
  • Surgical grade
  • Rockwell Hardness (HRC): 40–45
  • Fully autoclavable
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are compatible with a variety of sterilization methods, and are available in an assortment of lengths and jaw types.