VWR® Bone Nipper
Supplier: VWR
VWR®
11647-252
10806-284EA 225.66 CAD
10806-284
VWR® Bone Nipper
Bone Cutters and Rongeurs
The VWR® Bone Nipper is made from ASTM (899-94) Series 400 German stainless steel
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Designed for cutting cartilage and bone, this instrument features a strong, removable bar-spring that makes the shears self-opening.