VWR® Liston Bone Cutting Forceps
Supplier: VWR International
10806-268 10806-270 10806-272 10806-274 10806-276 10806-278
10806-268EA 484.22 CAD
10806-268 10806-270 10806-272 10806-274 10806-276 10806-278
VWR® Liston Bone Cutting Forceps
Bone Cutters and Rongeurs
The VWR® Liston Bone Cutting Forceps are made from ASTM (899-94) Series 400 German stainless steel
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in straight or curved formats, these instruments are designed for cutting cartilage and bone. A strong, removable bar-spring makes these shears self-opening.
Order Now
SPECIFICATIONS
Environmentally Preferable