VWR® Mini and Micro Friedman Rongeurs
Supplier: VWR International
10806-416 10806-418
10806-416EA 543.42 CAD
10806-416 10806-418
VWR® Mini and Micro Friedman Rongeurs
Bone Cutters and Rongeurs
Available in either Mini or Micro sizes, the VWR® Friedman Rongeurs are made from ASTM (899-94) Series 400 German stainless steel.
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Designed specifically for cutting cartilage and bone, these instruments feature a strong, removable bar-spring that makes the shears self-opening.