VWR® Cell Culture Dishes Treated for Increased Cell Attachment, Sterile
Supplier: VWR International
10861-656 10861-658 10861-660 10861-662 10861-680 10861-682
10861-656CS 483.55 CAD
10861-656 10861-658 10861-660 10861-662 10861-680 10861-682
VWR® Cell Culture Dishes Treated for Increased Cell Attachment, Sterile
Culture Dishes Cell Culture Dishes
Increased cell attachment surface treatment can improve cell spreading and attachment.
  • Available with 6 different diameters
  • Increased cell attachment surface treated
  • Flat bottom, uniform wall thickness ensures distortion-free bottom
  • Rim on the bottom side of the lid mates with the dish brim for easy and secure stacking
  • Numeric scale divides dish into quadrants for easy orientation
  • DNase- and RNase-free, and non-pyrogenic
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell cultureMore Product Information
Order Now
SPECIFICATIONS
Colour Clear
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M