Ethanol denatured, HiPerSolv CHROMANORM® for HPLC (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Ethyl alcohol denatured

BDH67002.400
BDH67002.400CS 1181.24 CAD
BDH67002.400
Ethanol denatured, HiPerSolv CHROMANORM® for HPLC (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Ethanol denatured
Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114 °C
Flash Pt: 12 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Ethyl Alcohol 89-91%
Isopropyl alcohol 4-6%
Methyl alcohol 4-6%
Color Max 10 APHA
Residue after evaporation Max 4 ppm
Water (by KF coulometry) Max 0.10%
Transmittance at 205 nm Min 10 T(%)
Transmittance at 210 nm Min 20 T(%)
Transmittance at 220 nm Min 45 T(%)
Transmittance at 230 nm Min 60 T(%)
Transmittance at 250 nm Min 90 T(%)
Transmittance at 270 nm Min 98 T(%)
Transmittance at 300 nm Min 99 T(%)
Optical absorbance at 205 nm Max 1.00 AU
Optical absorbance at 210 nm Max 0.65 AU
Optical absorbance at 220 nm Max 0.35 AU
Optical absorbance at 230 nm Max 0.20 AU
Optical absorbance at 250 nm Max 0.04 AU
Optical absorbance at 270 nm Max 0.01 AU
Optical absorbance at 300 nm Max 0.005 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR