Chloroform ≥99.90% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Methane, trichloro-TrichloroformMethylidyne trichlorideTrichloromethane

Stabilization: Stabilised with ethanol

BDH83627.400
BDH83627.400CS 1688.62 CAD
BDH83627.400
Chloroform ≥99.90% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://ca.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg

Order Now

Specification Test Results

Acetone and Aldehyde (as Acetone) Passes Test
Acid and chloride Passes Test
Assay (by GC-FID) (excluding ethanol) 99.90%
Color Max 10 APHA
Free chlorine Passes Test
Lead (Pb) Max 0.05 ppm
Residue after evaporation Max3 ppm
Stabilizer (as ethyl alcohol) 0.5-1.0%
Substances darkened by sulfuric acid Passes Test
Suitability for use in Dithizone tests Passes Test
Water (by KF coulometry) Max 0.02%
Transmittance at 245 nm Min 10 T(%)
Transmittance at 255 nm Min 70 T(%)
Transmittance at 260 nm Min 90 T(%)
Transmittance at 270 nm Min 95 T(%)
Transmittance at 290 nm Min 98 T(%)
Transmittance at 400 nm Min 98 T(%)
Optical absorbance at 245 nm Max 1.00 AU
Optical absorbance at 250 nm Max 0.15 AU
Optical absorbance at 260 nm Max 0.05 AU
Optical absorbance at 270 nm Max 0.02 AU
Optical absorbance at 290 nm Max 0.01 AU
Optical absorbance at 400 nm Max 0.01 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR