VWR® Tissue Culture Plates, Non-Treated, Sterilized, Non-Pyrogenic
Supplier: VWR International
10813-630 10814-224 10861-554 10861-556 10861-558 10861-560 10861-562 10861-564 75780-348
10813-630CS 541.61 CAD
10813-630 10814-224 10861-554 10861-556 10861-558 10861-560 10861-562 10861-564 75780-348
VWR® Tissue Culture Plates, Non-Treated, Sterilized, Non-Pyrogenic
Microplates Cell Culture Plates
Cell and tissue culture plates are ideal for cell growth and cell yields.
  • Available with 8 different growth surface areas
  • Uniform well volume
  • Flat well bottom and round bottom plates available
  • Well surface is smooth and free from striation
  • Alphanumerically coded for easy identification
  • Suitable for all common instruments and automation
  • Sterilized by electron beam irradiation
  • Non-pyrogenic
Raised rims on wells with the uniform rings on the lid help to reduce evaporation.
Plates are designed for use in applications where cell attachment is notMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M