TB Base Digestant, Hardy Diagnostics
Supplier: Hardy Diagnostics
U22 U20 U20
CA10766-744PK 326.59 CAD
CA10766-744 CA10001-504 10324-400
TB Base Digestant, Hardy Diagnostics
Cleaning Agents Detergents and Surface Cleansers
TB Base Digestant to be used with N-acetyl-L-cystiene for the processing of specimens for AFB culture.
  • Digests and decontaminates specimens
  • Process specimens for AFB culture
  • Plastic, screw cap bottle
TB Base Digestant to be used with N-acetyl-L-cystiene for the processing of specimens for AFB culture. Digests and decontaminates specimens prior to TB culture. Packaged in a plastic screw-cap bottle.