VWR® Nichrome/Aluminum Innoculating Loops
Supplier: VWR International
10806-216 10806-218 10806-220 10806-222 10806-224 10806-226 10806-228 10806-230 10806-232 10806-234 10806-236 10806-238 10806-354 10806-356 10806-358 10806-360
10806-216EA 29.35 CAD
10806-216 10806-218 10806-220 10806-222 10806-224 10806-226 10806-228 10806-230 10806-232 10806-234 10806-236 10806-238 10806-354 10806-356 10806-358 10806-360
VWR® Nichrome/Aluminum Innoculating Loops
Spreaders Inoculating Loops and Needles
VWR® Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle.
  • Passivated
  • 203 mm (8") handles, available with insulation
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The aluminum handle is available in either an insulated or non-insulated format. Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.