PYR Test Kit, with 15 ml Chromogenic Solution and Test Cards, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z275 Z275
CA10001-522EA 339.61 CAD
CA10001-522 10324-574
PYR Test Kit, with 15 ml Chromogenic Solution and Test Cards, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
PYR Test Kit and PYR Reagent are recommended for the detection of pyrolidonyl arylamidase activity.
  • Three tests can be performed on one card
  • Card format is easier to handle than disks
Hardy Diagnostics PYR Test Kit and PYR Reagent are recommended for the detection of pyrolidonyl arylamidase (also called pyrolidonyl aminopeptidase) activity in certain groups of bacteria, such as Streptococcus pyogenes (group A strep), Enterococcus spp., some coagulase-negative staphylococci, and some Enterobacteriaceae. The card format is easier to handle than disks. Three tests can be performed on one card.