Haemophilus ID Quadplate, for X and V Factors, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
J82 J82
CA10001-458PK 226.42 CAD
CA10001-458 10324-556
Haemophilus ID Quadplate, for X and V Factors, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Haemophilus ID Quadplate consists of four separate formulations
  • RTF Casman Medium
  • Modified (X- and V-factors)
  • Tryptic Soy Agar (TSA)
  • Chocolate Agar (X- and V-factors)
Haemophilus ID Quadplate consists of four separate formulations, which supply the requirements as follows: RTF Casman Medium, Modified (X- and V-factors); Tryptic Soy Agar (TSA) with Hemin (X-factor); Tryptic Soy Agar (TSA), modified with NAD (V-factor) ; Chocolate Agar (X- and V-factors).