KIMBLE® Open-Top Polypropylene Screw Thread Caps with Septa, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
N73805-24 73812BK-8425 73813BK-8425 73812WH-8425 73813WH-8425 73812BK-9425 73813BK-9425 73812BL-9425 73813BL-9425 73812BK-13425 73813BK-13425 73812WH-13425 73813WH-13425 73814WH-8425 73814BL-9425 73814BK-13425 73814BK-13425
93001-090CS 652.56 CAD
93001-090 89201-886 89201-888 89201-890 89201-892 89201-894 89201-896 89201-898 89201-900 89201-902 89201-904 89201-906 89201-908 89201-910 89201-912 89201-914 10061-466
KIMBLE® Open-Top Polypropylene Screw Thread Caps with Septa, DWK Lifesciences
Closures Test/Sample Tube Closures
Convenient pre-assembled caps and liners reduce the risk of contamination which makes this to use in autosamplers.
  • Intended for both single and multiple use injection
  • Ideal for storage of volatile compounds and chemicals
  • Autoclavable
Screw thread caps with bonded white PTFE septa available only as standard (no slit), whereas caps with red PTFE faced silicone septa available as both standard and pre slit.