BHI (Brain Heart Infusion) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G14 K258
CA10001-428PK 174.78 CAD
CA10001-428 CA10001-468
BHI (Brain Heart Infusion) Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
BHI Agar is used for cultivation, isolation, and selection of fungi.
  • Cultivation of microorganisms
  • Cultivation of fungi
  • Broth for cultivation of gram negative rods
For the cultivation of microorganisms. For the cultivation of fungi. Broth for cultivation of ciprofloxacin-resistant E. coli and other gram negative rods.