VWR® Dissection Needle with Plastic Holder
Supplier: VWR International
10806-326 10806-328
10806-326EA 18.43 CAD
10806-326 10806-328
VWR® Dissection Needle with Plastic Holder
Dissection Needles
Sharp-pointed, straight needles feature a plastic holder.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in straight or bent formats, all devices have a needle length of 45mm (1.75").