Middlebrook 7H11 Pour/ Selective Biplate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
J75 SP57
CA10001-456PK 209.73 CAD
CA10001-456 CA10001-498
Middlebrook 7H11 Pour/ Selective Biplate, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the isolation and detection of mycobacteria using the microcolony method.
  • Thin pour plate for microscopic examination
  • Isolation and detection of mycobacteria
  • Microcolony method
For the isolation and detection of mycobacteria using the microcolony method. Thin pour plate for microscopic examination. Use with MycoSeals. Biplate for For the cultivation of Mycobacterium spp.