Saline, 0.85%, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K59 R55 K52 K58
CA10001-472PK 147.57 CAD
CA10001-472 CA10001-494 CA77497-278 CA77497-258
Saline, 0.85%, Hardy Diagnostics
Buffers Cell Culture Buffers
For the preparation of isotonic suspensions and dilutions of microorganisms.