VWR® Flask Tongs
Supplier: VWR
VWR®
11647-161
10806-308EA 56.36 CAD
10806-308
VWR® Flask Tongs
Tongs
VWR® Flask Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
  • Passivated
  • "V" shaped jaws
  • Precision riveted joint
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Tongs are designed for the safe and simple handling of beakers and similar vessels ranging in size from 50 mL to 2000 mL. Precision Riveted Joint. Fiberglass sleeves aid in high-temperature applications.